Chemical Identifiers
CAS143491-57-0
IUPAC Name4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC8H10FN3O3S
InChI KeyXQSPYNMVSIKCOC-NTSWFWBYSA-N
SMILESNC1=NC(=O)N(C=C1F)[C@@H]1CS[C@H](CO)O1
View more