Thermo Scientific Chemicals

Isoprenaline hemisulfate

Catalog number: 462110250
25 g, Each
Thermo Scientific Chemicals

Isoprenaline hemisulfate

Catalog number: 462110250
25 g, Each
Quantity
Catalog number: 462110250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
299-95-6
IUPAC Name
bis(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol); sulfuric acid
Molecular Formula
C22H36N2O10S
InChI Key
ZOLBALGTFCCTJF-UHFFFAOYNA-N
SMILES
OS(O)(=O)=O.CC(C)NCC(O)C1=CC=C(O)C(O)=C1.CC(C)NCC(O)C1=CC=C(O)C(O)=C1
Water
=<7 %
HPLC
>=97.5 %
Identity
passes test
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text