Chemical Identifiers
CAS871038-72-1
IUPAC Namepotassium 4-{[(4-fluorophenyl)methyl]carbamoyl}-1-methyl-2-{2-[(5-methyl-1,3,4-oxadiazol-2-yl)formamido]propan-2-yl}-6-oxo-1,6-dihydropyrimidin-5-olate
Molecular FormulaC20H20FKN6O5
InChI KeyIFUKBHBISRAZTF-UHFFFAOYSA-M
SMILES[K+].CN1C(=O)C([O-])=C(N=C1C(C)(C)NC(=O)C1=NN=C(C)O1)C(=O)NCC1=CC=C(F)C=C1
View more