Saxagliptin, Thermo Scientific Chemicals
Saxagliptin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Saxagliptin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462180010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS361442-04-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only