Thermo Scientific Chemicals

Saxagliptin, Thermo Scientific Chemicals

Catalog number: 462182500
250 mg, Each
Thermo Scientific Chemicals

Saxagliptin, Thermo Scientific Chemicals

Catalog number: 462182500
250 mg, Each
Quantity
Catalog number: 462182500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
361442-04-8
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text