Sertraline hydrochloride
Sertraline hydrochloride
Sertraline hydrochloride
Sertraline hydrochloride
Thermo Scientific Chemicals

Sertraline hydrochloride

CAS: 79559-97-0 | C17H17Cl2N·HCl | 342.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462190010
Price (USD)
47.65
Online Exclusive
52.70
Save 5.05 (10%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
52.70
Save 5.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS79559-97-0
IUPAC Namehydrogen (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine chloride
Molecular FormulaC17H18Cl3N
InChI KeyBLFQGGGGFNSJKA-XHXSRVRCSA-N
SMILES[H+].[Cl-].CN[C@H]1CC[C@@H](C2=CC=C(Cl)C(Cl)=C2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Water=<0.5 %
HPLC>=97.5 %
Specific optical rotation+38° to +43° (20°C, 589 nm) (2 % in CH3OH)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link