Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Sibutramine hydrochloride monohydrate
Thermo Scientific Chemicals

Sibutramine hydrochloride monohydrate

CAS: 125494-59-9 | C17H29Cl2NO | 334.33 g/mol
Have Questions?
Catalog NumberQuantity
4622000101 g
Catalog number 462200010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS125494-59-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point194°C to 199°C
Water4.5 to 6 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0399467Certificate of AnalysisAug 25, 2023462200050, 462200010
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link