Tenax™ TA, 80-100 mesh, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

Tenax™ TA, 80-100 mesh, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Catalog number 462210100
Price (USD)/ Each
535.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
535.00
Add to cart
Tenax™ TA, 80-100 mesh, Thermo Scientific Chemicals
Catalog number462210100
Price (USD)/ Each
535.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-methoxy-5-methyl-3-phenyl-1,1'-biphenyl
Molecular FormulaC20H18O
InChI KeyTWADJGWUKGOPFG-UHFFFAOYSA-N
SMILESCOC1=C(C=C(C)C=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)274.36
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Particle size>=60 % (80/100 mesh)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only