Thermo Scientific Chemicals

Trimethyl citrate, Thermo Scientific Chemicals

Catalog number: 462250050
5 g, Each
Thermo Scientific Chemicals

Trimethyl citrate, Thermo Scientific Chemicals

Catalog number: 462250050
5 g, Each
Quantity
Catalog number: 462250050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1587-20-8
IUPAC Name
1,2,3-trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C9H14O7
InChI Key
HDDLVZWGOPWKFW-UHFFFAOYSA-N
SMILES
COC(=O)CC(O)(CC(=O)OC)C(=O)OC
Appearance (Form)
Crystals
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Melting point
75°C to 82°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text