Thermo Scientific Chemicals

Vindoline

Catalog number: 462270050
5 g, Each
Thermo Scientific Chemicals

Vindoline

Catalog number: 462270050
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 462270050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2182-14-1
IUPAC Name
methyl (1R,9R,11R,12R,19R)-11-(acetyloxy)-12-ethyl-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2,4,6,13-tetraene-10-carboxylate
Molecular Formula
C25H32N2O6
InChI Key
CXBGOBGJHGGWIE-MXXIQHGTSA-N
SMILES
CC[C@@]12C=CCN3CC[C@@]4([C@H]13)[C@@H](N(C)C1=CC(OC)=CC=C41)C(O)([C@@H]2OC(C)=O)C(=O)OC
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Loss on drying
=<1 %
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text