Thermo Scientific Chemicals

7-Hydroxyindole, 95%, Thermo Scientific Chemicals

Catalog number: 462292500
250 mg, Each
Thermo Scientific Chemicals

7-Hydroxyindole, 95%, Thermo Scientific Chemicals

Catalog number: 462292500
250 mg, Each
Quantity
Catalog number: 462292500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2380-84-9
IUPAC Name
1H-indol-7-ol
Molecular Formula
C8H7NO
InChI Key
ORVPXPKEZLTMNW-UHFFFAOYSA-N
SMILES
OC1=C2NC=CC2=CC=C1
Appearance (Color)
Yellow to brown
Appearance (Form)
Solid or crystalline solid
Infrared spectrum
Conforms
Melting point
83°C to 88°C
GC
>=94.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text