Thermo Scientific Chemicals

Citalopram hydrobromide

Catalog number: 462310010
1 g, Each
Thermo Scientific Chemicals

Citalopram hydrobromide

Catalog number: 462310010
1 g, Each
Quantity
Catalog number: 462310010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59729-32-7
IUPAC Name
hydrogen 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile bromide
Molecular Formula
C20H22BrFN2O
InChI Key
WIHMBLDNRMIGDW-UHFFFAOYNA-N
SMILES
[H+].[Br-].CN(C)CCCC1(OCC2=CC(=CC=C12)C#N)C1=CC=C(F)C=C1
Loss on drying
=<0.5 % (1 g, 105°C, 4 h)
HPLC
>=97.5 %
Solubility
(1 % in ethanol) Clear colorless
Appearance (Color)
White to almost white
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text