Thermo Scientific Chemicals

Erlotinib hydrochloride

Catalog number: 462330050
5 g, Each
Thermo Scientific Chemicals

Erlotinib hydrochloride

Catalog number: 462330050
5 g, Each
Quantity
Catalog number: 462330050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
183319-69-9
IUPAC Name
N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
Molecular Formula
C22H24ClN3O4
InChI Key
GTTBEUCJPZQMDZ-UHFFFAOYSA-N
SMILES
Cl.COCCOC1=C(OCCOC)C=C2C(NC3=CC=CC(=C3)C#C)=NC=NC2=C1
Appearance (Color)
White to yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Solubility
(1% in DMSO) Clear solution
Loss on drying
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text