Propafenone hydrochloride
Product Image
Thermo Scientific Chemicals

Propafenone hydrochloride

CAS: 34183-22-7 | C21H28ClNO3 | 377.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462390050
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS34183-22-7
IUPAC Namehydrogen 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one chloride
Molecular FormulaC21H28ClNO3
InChI KeyXWIHRGFIPXWGEF-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder
Loss on drying=<0.5 %
pH5.0 to 6.2 (0.5 % in water)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection