1x1 image pixel for data collection
Granisetron hydrochloride, 98%
Granisetron hydrochloride, 98%
Thermo Scientific Chemicals

Granisetron hydrochloride, 98%

CAS: 107007-99-8 | C18H25ClN4O | 348.88 g/mol
Have Questions?
Catalog NumberQuantity
4624800101 g
Catalog number 462480010
Price (USD)
245.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
245.00
Each
Add to cart
Chemical Identifiers
CAS107007-99-8
IUPAC Namehydrogen 1-methyl-N-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-1H-indazole-3-carboxamide chloride
Molecular FormulaC18H25ClN4O
InChI KeyQYZRTBKYBJRGJB-UHFFFAOYNA-N
SMILES[H+].[Cl-].CN1N=C(C(=O)NC2CC3CCCC(C2)N3C)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Solubility(1 % in water) Clear colorless solution
Loss on drying=<0.5 %
Appearance (Color)White to almost white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link