Labetalol hydrochloride, 99%
Thermo Scientific Chemicals
Labetalol hydrochloride, 99%
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 462600050
Price (USD)/ Each
106.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
106.00
Add to cart
Labetalol hydrochloride, 99%
Catalog number462600050
Price (USD)/ Each
106.00
-
Add to cart
Chemical Identifiers
CAS32780-64-6
IUPAC Name2-hydroxy-5-{1-hydroxy-2-[(4-phenylbutan-2-yl)amino]ethyl}benzamide hydrochloride
Molecular FormulaC19H25ClN2O3
InChI KeyWQVZLXWQESQGIF-UHFFFAOYNA-N
SMILESCl.CC(CCC1=CC=CC=C1)NCC(O)C1=CC=C(O)C(=C1)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Melting point178°C to 180°C
Appearance (Form)Powder
Loss on drying=<1 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only