Eflornithine hydrochloride monohydrate
Product Image
Thermo Scientific Chemicals

Eflornithine hydrochloride monohydrate

CAS: 96020-91-6 | C6H15ClF2N2O3 | 236.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number 462700250
Price (USD)
407.65
Online Exclusive
453.00
Save 45.35 (10%)
Each
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
407.65
Online Exclusive
453.00
Save 45.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS96020-91-6
IUPAC Namehydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
Molecular FormulaC6H15ClF2N2O3
InChI KeyFJPAMFNRCFEGSD-UHFFFAOYNA-N
SMILES[H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Water6 to 9 %
Titration with HClO4>=97.5 % (dry substance)
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0385296Certificate of AnalysisMay 17, 2025462700250, 462700050, 462700010
A0385296Certificate of AnalysisSep 05, 2023462700250, 462700050, 462700010
2 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection