Thermo Scientific Chemicals

(S,R)-Noscapine

Catalog number: 462710250
25 g, Each
Thermo Scientific Chemicals

(S,R)-Noscapine

Catalog number: 462710250
25 g, Each
Quantity
Catalog number: 462710250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128-62-1
Appearance (Color)
White to almost white
Appearance (Color)
Crystalline powder
Infrared spectrum
Conforms
Melting point
174°C to 177°C
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text