Thermo Scientific Chemicals

Palonosetron hydrochloride, Thermo Scientific Chemicals

Catalog number: 462990010
1 g, Each
Thermo Scientific Chemicals

Palonosetron hydrochloride, Thermo Scientific Chemicals

Catalog number: 462990010
1 g, Each
Quantity
Catalog number: 462990010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
135729-62-3
IUPAC Name
hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),9,11-trien-2-one chloride
Molecular Formula
C19H25ClN2O
InChI Key
OLDRWYVIKMSFFB-KPVRICSOSA-N
SMILES
[H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2
HPLC
>=98.5 %
Loss on drying
=<1 % (80°C, 3 h) (vacuum)
Residue after ignition
=<0.1 %
Identification
Passes test for Chlorides
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text