Altretamine
Thermo Scientific Chemicals

Altretamine

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 463090010
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
Add to cart
Altretamine
Catalog number463090010
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS645-05-6
IUPAC NameN2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine
Molecular FormulaC9H18N6
InChI KeyUUVWYPNAQBNQJQ-UHFFFAOYSA-N
SMILESCN(C)C1=NC(=NC(=N1)N(C)C)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<1 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only