Thermo Scientific Chemicals

Repaglinide

Catalog number: 463232500
250 mg, Each
Thermo Scientific Chemicals

Repaglinide

Catalog number: 463232500
250 mg, Each
Quantity
Catalog number: 463232500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
135062-02-1
IUPAC Name
2-ethoxy-4-({[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]carbamoyl}methyl)benzoic acid
Molecular Formula
C27H36N2O4
InChI Key
FAEKWTJYAYMJKF-QHCPKHFHSA-N
SMILES
CCOC1=CC(CC(=O)N[C@@H](CC(C)C)C2=CC=CC=C2N2CCCCC2)=CC=C1C(O)=O
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (105°C)
Specific optical rotation
+7° to +10° (20°C, 589 nm) (1% in MeOH)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text