1x1 image pixel for data collection
Product Image
Thermo Scientific Chemicals

Tolcapone

Have Questions?
Catalog NumberQuantity
463282500250 mg
Catalog number 463282500
Price (USD)
168.00
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
168.00
Each
Add to cart
Chemical Identifiers
CAS134308-13-7
IUPAC Name5-(4-methylbenzoyl)-3-nitrobenzene-1,2-diol
Molecular FormulaC14H11NO5
InChI KeyMIQPIUSUKVNLNT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C1=CC(O)=C(O)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder3
Infrared spectrumConforms
HPLC>=96.0 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link

1x1 image pixel for data collection