Sodium sulfobutyl ether beta-cyclodextrin, Thermo Scientific Chemicals
Sodium sulfobutyl ether beta-cyclodextrin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfobutyl ether beta-cyclodextrin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 463360050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS182410-00-0
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
pH4.0 to 6.8 (30% in water)
Solubility(30% in water) Clear solution
Appearance (Color)White to almost white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only