Product Image
Thermo Scientific Chemicals

Etoricoxib

CAS: 202409-33-4 | C18H15ClN2O2S | 358.84 g/mol
Have Questions?
Catalog NumberQuantity
4633800101 g
Catalog number 463380010
Price (USD)
133.65
Special offer
Online exclusive
157.00
Save 23.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
133.65
Special offer
Online exclusive
157.00
Save 23.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS202409-33-4
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Appearance (Form)Crsytalline powder
Infrared spectrumConforms
Appearance (Color)White to light yellow
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link