Product Image
Thermo Scientific Chemicals

(±)-Sulpiride

CAS: 15676-16-1 | C15H23N3O4S | 341.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 463480250
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS15676-16-1
IUPAC NameN-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
Molecular FormulaC15H23N3O4S
InChI KeyBGRJTUBHPOOWDU-UHFFFAOYNA-N
SMILESCCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after ignition=<0.1 %
Appearance (Form)Powder and or crystals
HPLC>=97.5 %
Loss on drying=<0.5 % (1 g, 105°C)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link