Thermo Scientific Chemicals

Ethano L, 99.8%, anhyd.,denat. with 1%iPr-OH,1%MEK,0.001%Bitrex(R), Thermo Scientific Chemicals

Catalog number: 463680010
1 L, Each
Thermo Scientific Chemicals

Ethano L, 99.8%, anhyd.,denat. with 1%iPr-OH,1%MEK,0.001%Bitrex(R), Thermo Scientific Chemicals

Catalog number: 463680010
1 L, Each
Quantity
Catalog number: 463680010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
Appearance (Color)
Clear colorless
Refractive index
1.3610 to 1.3630 (20°C, 589 nm)
Water
=<3400 ppm
Density
(20°C) 0.789 to 0.790 kg/l
Ethanol
99.8 to 100.0 % v/v (calculated)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text