Ethanol, 96%, specified according to the requirements of Ph. Eur, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
Ethanol, 96%, specified according to the requirements of Ph. Eur, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
Thermo Scientific Chemicals

Ethanol, 96%, specified according to the requirements of Ph. Eur, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™

CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463690250
Price (EUR)
-
Quantity:
25 L
Request bulk or custom format
Chemical Identifiers
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
Molecular Weight (g/mol)46.07
View more
SpecificationsSpecification SheetSpecification Sheet
DenaturantDenatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use

Share catalog number, name or link