Ethanol, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
Ethanol, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
Thermo Scientific Chemicals

Ethanol, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™

CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463700250
Price (USD)
-
Quantity:
25 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
DenaturantDenatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0454665Certificate of AnalysisAug 25, 2023463700025, 463700050, 463700250
A0455568Certificate of AnalysisAug 25, 2023463700025, 463700050, 463700250
A0449788Certificate of AnalysisFeb 02, 2023463700025, 463700050, 463700250
A0448739Certificate of AnalysisJan 06, 2023463700025, 463700050, 463700250
A0411875Certificate of AnalysisDec 13, 2022463700025, 463700050, 463700250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection