Thermo Scientific Chemicals

10H-Phenothiazine-10-propanesulfonic acid sodium salt, Thermo Scientific Chemicals

Catalog number: 464000050
5 g, Each
Thermo Scientific Chemicals

10H-Phenothiazine-10-propanesulfonic acid sodium salt, Thermo Scientific Chemicals

Catalog number: 464000050
5 g, Each
Quantity
Catalog number: 464000050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101199-38-6
IUPAC Name
sodium 3-(10H-phenothiazin-10-yl)propane-1-sulfonate
Molecular Formula
C15H14NNaO3S2
InChI Key
VKGYKXFNSKEHED-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)CCCN1C2=CC=CC=C2SC2=CC=CC=C12
Appearance (Color)
Almost white to grey to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text