Methanol-d4,, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

Methanol-d4,, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
10 mL
25 mL
50 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464040100
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
-
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
Add to cart
Methanol-d4,, Thermo Scientific Chemicals
Catalog number464040100
Price (USD)/ Each
96.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
113.00 
Save 16.35 (14%)
-
Add to cart
Chemical Identifiers
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)liquid
Water=<0.03 % (Coulometric)
Appearance (Color)Clear colorless
D-Enrichment>=99.75 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only