Thermo Scientific Chemicals

Acetic acid-d, for NMR, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464150250
25 mL, Each
Thermo Scientific Chemicals

Acetic acid-d, for NMR, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464150250
25 mL, Each
Quantity
Catalog number: 464150250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
758-12-3
IUPAC Name
acetate
Molecular Formula
C2H3O2
InChI Key
QTBSBXVTEAMEQO-UHFFFAOYSA-M
SMILES
CC([O-])=O
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.4 %
Water
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text