Acetonitrile-d3, for NMR, 100 atom % D, packaged in 0.75 ml ampoules, Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, 100 atom % D, packaged in 0.75 ml ampoules, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, 100 atom % D, packaged in 0.75 ml ampoules, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
7.5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464170075
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
7.5 mL
Request bulk or custom format
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
Add to cart
Acetonitrile-d3, for NMR, 100 atom % D, packaged in 0.75 ml ampoules, Thermo Scientific Chemicals
Catalog number464170075
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2206-26-0
IUPAC Name(²H₃)acetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.95 %
Water=<0.02 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only