Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464210075
7.5 mL, Each
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464210075
7.5 mL, Each
Quantity
Catalog number: 464210075
Price (USD)
Have Questions?

Chemical Identifiers

CAS
33685-54-0
IUPAC Name
tetrachloro(²H₂)ethane
Molecular Formula
C2H2Cl4
InChI Key
QPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES
[2H]C(Cl)(Cl)C([2H])(Cl)Cl
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.4 %
Water
=<0.03 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text