Thermo Scientific Chemicals

Trifluoroacetic acid-d, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464220075
7.5 mL, Each
Thermo Scientific Chemicals

Trifluoroacetic acid-d, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 464220075
7.5 mL, Each
Quantity
Catalog number: 464220075
Price (USD)
Have Questions?

Chemical Identifiers

CAS
599-00-8
IUPAC Name
trifluoroethan(²H)oic acid
Molecular Formula
C2HF3O2
InChI Key
DTQVDTLACAAQTR-DYCDLGHISA-N
SMILES
[2H]OC(=O)C(F)(F)F
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.4 %
Water
=<0.05 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text