Thermo Scientific™

Restore™ PLUS Western Blot Stripping Buffer

Catalog number: 46428
Have Questions?
Thermo Scientific™

Restore™ PLUS Western Blot Stripping Buffer

Catalog number: 46428
Have Questions?
Catalog Number
46428
Unit Size
30 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4642830 mL
-
46430X44 x 500 mL
-
46430500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Restore PLUS Western Blot Stripping Buffer is an advanced formula for removing bound primary and secondary antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.

Features of Restore PLUS Western Blot Stripping Buffer:

Ready and easy to use—no dilution necessary; no offensive odors; store at room temperature
Compatible—use on nitrocellulose and PVDF membranes, whether still wet or already dry; works with practically any blocking buffer, enzyme conjugate and chemiluminescent substrate
Cost effective—save valuable time and samples; strip blots effectively the first time
Robust yet gentle—transferred proteins remain viable; strip the same blot up to five times
Flexible—strip and reprobe to optimize antibody concentrations or to detect a new antigen with different antibodies

Restore PLUS Buffer is an alternative formulation of the original Thermo Scientific Restore Western Blot Stripping Buffer. Restore PLUS Stripping Buffer was designed for use with antibodies that are difficult to remove from western blots and require longer incubation times or incubation temperatures greater than 22°C with gentler formulations. High-affinity antibodies can be quickly and effectively stripped from western blots at room temperature without removing transferred proteins, thereby allowing multiple reprobes of the target.

Protocol Summary:
• Wash blot to remove chemiluminescent substrate
• Incubate blot in Restore Western Blot Stripping Buffer for 5 to 15 minutes at room temperature
(or incubate at 37°C for high affinity antibodies)
• Remove blot and wash in Wash Buffer
• Block membrane
• Test for sufficient removal of antibodies
• Perform next immunoblot experiment
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Western Blot Stripping Buffer
Detection Method
Chemiluminescent
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
Restore™
Quantity
30 mL

Contents & Storage

Upon receipt store at room temperature. Product shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text