Chemical Identifiers
CAS5786-21-0
IUPAC Name6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
Molecular FormulaC18H19ClN4
InChI KeyZUXABONWMNSFBN-UHFFFAOYSA-N
SMILESCN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
View more