Carteolol hydrochloride
Product Image
Thermo Scientific Chemicals

Carteolol hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464320050
Price (USD)
377.65
Online Exclusive
419.00
Save 41.35 (10%)
Each
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
377.65
Online Exclusive
419.00
Save 41.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS51781-21-6
IUPAC Namehydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
Molecular FormulaC16H25ClN2O3
InChI KeyFYBXRCFPOTXTJF-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Loss on drying=<0.5 % (1 g, 105°C)
Solubility(0.3 g in 10 ml water) Clear colorless solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link