Product Image
Thermo Scientific Chemicals

Vandetanib

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 464332500
Price (USD)/ Each
94.60
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
94.60
Add to cart
Vandetanib
Catalog number464332500
Price (USD)/ Each
94.60
-
Add to cart
Chemical Identifiers
CAS443913-73-3
IUPAC NameN-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Molecular FormulaC22H24BrFN4O2
InChI KeyUHTHHESEBZOYNR-UHFFFAOYSA-N
SMILESCOC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<0.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only