Thermo Scientific Chemicals

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals

Catalog number: 464350010
1 g, Each
Thermo Scientific Chemicals

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals

Catalog number: 464350010
1 g, Each
Quantity
Catalog number: 464350010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
947725-04-4
IUPAC Name
5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
Molecular Formula
C12H17F3S
InChI Key
VRTQPEYVMHATOA-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C
Appearance (Color)
White to light yellow to light red
Infrared spectrum
Conforms
Melting point
62°C to 68°C
Appearance (Color)
to light brown
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text