Hydrogen peroxide, 12% w/v, Thermo Scientific Chemicals
Hydrogen peroxide, 12% w/v, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen peroxide, 12% w/v, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number 464570010
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
Add to cart
Hydrogen peroxide, 12% w/v, Thermo Scientific Chemicals
Catalog number464570010
Price (USD)/ Each
38.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
45.30 
Save 6.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
Molecular Weight (g/mol)34.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration with KMnO411.0 to 13.0 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only