Chemical Identifiers
CAS2244386-63-6
IUPAC Namesodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular FormulaC23H35NaO7
InChI KeyVWBQYTRBTXKKOG-IYNICTALSA-M
SMILES[Na+].[H][C@]12[C@H](C[C@H](O)C=C1C=C[C@H](C)[C@@H]2CC[C@@H](O)C[C@@H](O)CC([O-])=O)OC(=O)[C@@H](C)CC
View more