Thermo Scientific Chemicals

Pravastatin sodium salt hydrate

Catalog number: 464882500
250 mg, Each
Thermo Scientific Chemicals

Pravastatin sodium salt hydrate

Catalog number: 464882500
250 mg, Each
Quantity
Catalog number: 464882500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2244386-63-6
IUPAC Name
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molecular Formula
C23H35NaO7
InChI Key
VWBQYTRBTXKKOG-IYNICTALSA-M
SMILES
[Na+].[H][C@]12[C@H](C[C@H](O)C=C1C=C[C@H](C)[C@@H]2CC[C@@H](O)C[C@@H](O)CC([O-])=O)OC(=O)[C@@H](C)CC
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Specific optical rotation
+150° to +159° (20°C, 589 nm) (0.5% in water)
Water
=<4 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text