Fexofenadine hydrochloride
Product Image
Thermo Scientific Chemicals

Fexofenadine hydrochloride

CAS: 153439-40-8 | C32H40ClNO4 | 538.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464972500
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS153439-40-8
IUPAC Namehydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
Molecular FormulaC32H40ClNO4
InChI KeyRRJFVPUCXDGFJB-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Form)Powder
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link