Thermo Scientific™
F1-ClipTip™ Fixed Volume Single Channel Pipettes
Catalog number: 4651220N
Have Questions?
Thermo Scientific™
F1-ClipTip™ Fixed Volume Single Channel Pipettes
Catalog number: 4651220N
Have Questions?
With the added confidence of ClipTip technology, these pipettes are ideal for use in set protocols requiring a specific volume while providing low attachment and ejection forces.
Catalog Number
4651220N
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
50 μL
Compatible Tips
ClipTip 50
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 50
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)50 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
20 μL
Compatible Tips
ClipTip 20
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 20
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)20 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
25 μL
Compatible Tips
ClipTip 50
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 50
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)25 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
100 μL
Compatible Tips
ClipTip 200
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 200
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)100 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
200 μL
Compatible Tips
ClipTip 200
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 200
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)200 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
250 μL
Compatible Tips
ClipTip 300
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 300
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)250 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
500 μL
Compatible Tips
ClipTip 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 1000
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)500 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip Fixed Volume
Volume (Metric)
1000 μL
Compatible Tips
ClipTip 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 1000
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1000 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
F1-ClipTip
Volume (Metric)
10 μL
Compatible Tips
ClipTip 12.5 Ext
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsClipTip 12.5 Ext
DescriptionF1-ClipTip
ColorPink
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)10 μL
Increments0.02μL
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionVolume (Metric)Compatible TipsPrice (USD)AvailabilityQuantity
4651220NFull specifications
EachF1-ClipTip Fixed Volume50 μLClipTip 50
-
Compatible TipsClipTip 50
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)50 μL
Unit SizeEach
4651200NFull specifications
EachF1-ClipTip Fixed Volume20 μLClipTip 20
-
Compatible TipsClipTip 20
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)20 μL
Unit SizeEach
4651210NFull specifications
EachF1-ClipTip Fixed Volume25 μLClipTip 50
-
Compatible TipsClipTip 50
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)25 μL
Unit SizeEach
4651230NFull specifications
EachF1-ClipTip Fixed Volume100 μLClipTip 200
-
Compatible TipsClipTip 200
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)100 μL
Unit SizeEach
4651240NFull specifications
EachF1-ClipTip Fixed Volume200 μLClipTip 200
-
Compatible TipsClipTip 200
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)200 μL
Unit SizeEach
4651250NFull specifications
EachF1-ClipTip Fixed Volume250 μLClipTip 300
-
Compatible TipsClipTip 300
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)250 μL
Unit SizeEach
4651260NFull specifications
EachF1-ClipTip Fixed Volume500 μLClipTip 1000
-
Compatible TipsClipTip 1000
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)500 μL
Unit SizeEach
4651270NFull specifications
EachF1-ClipTip Fixed Volume1000 μLClipTip 1000
-
Compatible TipsClipTip 1000
DescriptionF1-ClipTip Fixed Volume
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1000 μL
Unit SizeEach
4651280NFull specifications
EachF1-ClipTip10 μLClipTip 12.5 Ext
-
Compatible TipsClipTip 12.5 Ext
DescriptionF1-ClipTip
ColorPink
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)10 μL
Increments0.02μL
Unit SizeEach
Showing 9 of 9
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research.
Learn more >
Ergonomically in-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries 
Secure volume lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable comfort 
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications. 
Wide selection of ClipTip Pipette Tips 
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time warranty
With convenient online registration, and may vary by country.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov