Aprepitant
Thermo Scientific Chemicals

Aprepitant

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465310010
Price (USD)/ Each
487.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
574.00 
Save 86.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
487.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
574.00 
Save 86.35 (15%)
Add to cart
Aprepitant
Catalog number465310010
Price (USD)/ Each
487.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
574.00 
Save 86.35 (15%)
-
Add to cart
Chemical Identifiers
CAS170729-80-3
IUPAC Name5-{[(2S,3R)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl}-2,3-dihydro-1H-1,2,4-triazol-3-one
Molecular FormulaC23H21F7N4O3
InChI KeyATALOFNDEOCMKK-BYYRLHKVSA-N
SMILESC[C@@H](O[C@@H]1OCCN(CC2=NC(=O)NN2)[C@@H]1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point250°C to 255°C
Specific optical rotation+66° to +71° (20°C, 589 nm) (1% in MeOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only