Thermo Scientific Chemicals

Travoprost, Thermo Scientific Chemicals

Catalog number: 465361000
100 mg, Each
Thermo Scientific Chemicals

Travoprost, Thermo Scientific Chemicals

Catalog number: 465361000
100 mg, Each
Quantity
Catalog number: 465361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
157283-68-6
IUPAC Name
propan-2-yl (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3R)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-en-1-yl]cyclopentyl]hept-5-enoate
Molecular Formula
C26H35F3O6
InChI Key
MKPLKVHSHYCHOC-AHTXBMBWSA-N
SMILES
CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)COC1=CC=CC(=C1)C(F)(F)F
Appearance (Color)
Colorless to light yellow
HPLC
>=95.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text