Chloroprocaine hydrochloride
Product Image
Thermo Scientific Chemicals

Chloroprocaine hydrochloride

Quantity:
250 mg
1 g
Catalog number 465370010
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3858-89-7
IUPAC Name2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride
Molecular FormulaC13H20Cl2N2O2
InChI KeySZKQYDBPUCZLRX-UHFFFAOYSA-N
SMILESCl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point173°C to 176°C
HPLC>=97.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection