1x1 image pixel for data collection
Chloroprocaine hydrochloride
Product Image
Thermo Scientific Chemicals

Chloroprocaine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465372500
Price (USD)
187.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
187.00
Each
Add to cart
Chemical Identifiers
CAS3858-89-7
IUPAC Name2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride
Molecular FormulaC13H20Cl2N2O2
InChI KeySZKQYDBPUCZLRX-UHFFFAOYSA-N
SMILESCl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point173°C to 176°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link