Thermo Scientific Chemicals

2,2'-Biimidazole, Thermo Scientific Chemicals

Catalog number: 465820050
5 g, Each
Thermo Scientific Chemicals

2,2'-Biimidazole, Thermo Scientific Chemicals

Catalog number: 465820050
5 g, Each
Quantity
Catalog number: 465820050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
492-98-8
IUPAC Name
1H,1'H-2,2'-biimidazole
Molecular Formula
C6H6N4
InChI Key
AZUHIVLOSAPWDM-UHFFFAOYSA-N
SMILES
N1C=CN=C1C1=NC=CN1
Appearance (Color)
Light yellow to brown to grey
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=97.5 %
Titration with HClO4
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text