Thermo Scientific Chemicals

Hexaethylene glycol monobenzyl ether, Thermo Scientific Chemicals

Catalog number: 465840010
1 g, Each
Thermo Scientific Chemicals

Hexaethylene glycol monobenzyl ether, Thermo Scientific Chemicals

Catalog number: 465840010
1 g, Each
Quantity
Catalog number: 465840010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24342-68-5
IUPAC Name
1-phenyl-2,5,8,11,14,17-hexaoxanonadecan-19-ol
Molecular Formula
C19H32O7
InChI Key
VVBQKDDPSXBMMZ-UHFFFAOYSA-N
SMILES
OCCOCCOCCOCCOCCOCCOCC1=CC=CC=C1
Refractive index
1.4920 to 1.4960 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=94 %
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text